missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2,3-dihydrobenzo[b]furan, 97%, Thermo Scientific™

Product Code. 11469756
Click to view available options
Quantity:
0.5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90843-31-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00068033
InChI Key MMVUJVASBDVNGJ-UHFFFAOYSA-N
Synonym 5-acetyl-2,3-dihydrobenzo b furan, 5-acetyl-2,3-dihydrobenzofuran, 1-2,3-dihydro-1-benzofuran-5-yl ethanone, 1-2,3-dihydrobenzofuran-5-yl ethanone, 1-2,3-dihydrobenzo b furan-5-yl ethan-1-one, 1-2,3-dihydro-benzofuran-5-yl-ethanone, 1-2,3-dihydro-5-benzofuranyl ethanone, 1-2,3-dihydro-1-benzofuran-5-yl ethan-1-one, 5-acetyl-2,3-dihydro-1-benzofuran
PubChem CID 145220
IUPAC Name 1-(2,3-dihydro-1-benzofuran-5-yl)ethanone
SMILES CC(=O)C1=CC=C2OCCC2=C1

Specifications

Melting Point 56°C to 59°C
Boiling Point 120°C to 121°C (15 mmHg)
Quantity 10 g
Beilstein 131198
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 5-Acetyl-2,3-dihydrobenzo[b]furan
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'