missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2,4-dimethylthiazole, 99%

Product Code. 11434774
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11434774

Brand: Thermo Scientific Alfa Aesar A13741.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38205-60-6 | C7H9NOS | 155.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38205-60-6
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD00005331
InChI Key BLQOKWQUTLNKON-UHFFFAOYSA-N
Synonym 5-acetyl-2,4-dimethylthiazole, 2,4-dimethyl-5-acetylthiazole, 1-2,4-dimethylthiazol-5-yl ethanone, 1-2,4-dimethyl-1,3-thiazol-5-yl ethanone, unii-pd36fu2wgy, ethanone, 1-2,4-dimethyl-5-thiazolyl, 1-2,4-dimethyl-5-thiazolyl ethanone, pd36fu2wgy, 2,4-dimethyl-5-thiazoyl methyl ketone, ketone, 2,4-dimethyl-5-thiazolyl methyl
PubChem CID 520888
IUPAC Name 1-(2,4-dimethyl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C)C(=O)C

Specifications

Density 1.15
Boiling Point 230°C
Flash Point 104°C (219°F)
Refractive Index 1.543
Quantity 25 g
Beilstein 120560
Formula Weight 155.22
Percent Purity 99%
Chemical Name or Material 5-Acetyl-2,4-dimethylthiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.