missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-methoxypyridine, 97%, Thermo Scientific Chemicals

Product Code. 11986381
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213193-32-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
InChI Key RYOQZXOVBJIUSX-UHFFFAOYSA-N
Synonym 5-acetyl-2-methoxypyridine, 1-6-methoxypyridin-3-yl ethanone, 1-6-methoxypyridin-3-yl ethan-1-one, 1-6-methoxy-3-pyridyl ethanone, ethanone, 1-6-methoxy-3-pyridinyl, 3-acetyl-6-methoxypyridine, 1-6-methoxy-3-pyridinyl ethanone, pubchem16770, acmc-209fj4, 2-methoxy-5-acetylpyridine
PubChem CID 10534837
IUPAC Name 1-(6-methoxypyridin-3-yl)ethanone
SMILES CC(=O)C1=CN=C(C=C1)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 53.0°C to 54.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 151.17
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 5-Acetyl-2-methoxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Acetyl-2-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'