missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Acetylthiophene-2-carbonitrile, 97%, Thermo Scientific™
£98.90 - £404.00
Chemical Identifiers
CAS | 88653-55-8 |
---|---|
Molecular Formula | C7H5NOS |
Molecular Weight (g/mol) | 151.183 |
InChI Key | VSHPLUBHIUFLES-UHFFFAOYSA-N |
Synonym | 2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ciShow More |
PubChem CID | 2747565 |
IUPAC Name | 5-acetylthiophene-2-carbonitrile |
SMILES | CC(=O)C1=CC=C(S1)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10612665
|
Thermo Scientific Maybridge
TL00354DE |
5 g | Amber Glass Bottle |
£98.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10556423
|
Thermo Scientific Maybridge
TL00354EA |
10 g | Amber Glass Bottle |
£182.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236004
|
Thermo Scientific Maybridge
TL00354EB |
25 g | Amber Glass Bottle |
£404.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
88653-55-8 | |
151.183 | |
2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ciShow More | |
5-acetylthiophene-2-carbonitrile |
C7H5NOS | |
VSHPLUBHIUFLES-UHFFFAOYSA-N | |
2747565 | |
CC(=O)C1=CC=C(S1)C#N |
Specifications
88653-55-8 | |
C7H5NOS | |
VSHPLUBHIUFLES-UHFFFAOYSA-N | |
5-acetylthiophene-2-carbonitrile | |
2747565 | |
97% |
97% | |
2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ci | |
CC(=O)C1=CC=C(S1)C#N | |
151.183 | |
151.19 | |
5-Acetylthiophene-2-carbonitrile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Acetylthiophene-2-carbonitrile, 97%, Thermo Scientific™