Learn More
5-Amino-1,3,4-thiadiazole-2-thiol, 98%, Thermo Scientific Chemicals
£21.41 - £108.54
Chemical Identifiers
CAS | 2349-67-9 |
---|---|
Molecular Formula | C2H3N3S2 |
Molecular Weight (g/mol) | 133.2 |
MDL Number | MFCD00003108 |
InChI Key | GDGIVSREGUOIJZ-UHFFFAOYSA-N |
Synonym | 5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole |
PubChem CID | 2723847 |
IUPAC Name | 5-amino-3H-1,3,4-thiadiazole-2-thione |
SMILES | C1(=NNC(=S)S1)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10476990
|
Thermo Scientific Acros
155970250 |
25 g | Glass bottle |
£21.41
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10532941
|
Thermo Scientific Acros
155970500 |
50 g | Glass bottle |
£30.95
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607322
|
Thermo Scientific Acros
155972500 |
250 g | Glass bottle |
£108.54
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2349-67-9 | |
133.2 | |
GDGIVSREGUOIJZ-UHFFFAOYSA-N | |
2723847 | |
C1(=NNC(=S)S1)N |
C2H3N3S2 | |
MFCD00003108 | |
5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole | |
5-amino-3H-1,3,4-thiadiazole-2-thione |
Specifications
2349-67-9 | |
100.0 | |
Cream to Yellow or White | |
Authentic | |
C2H3N3S2 | |
27, 674 | |
Solubility in water: insoluble. Other solubilities: soluble in nh4oh,soluble in dmf | |
C1(=NNC(=S)S1)N | |
133.2 | |
133.2 | |
Powder |
97.5 | |
231°C to 236°C | |
1% max. (110°C) | |
97.5% min. (HPLC) | |
MFCD00003108 | |
5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole | |
GDGIVSREGUOIJZ-UHFFFAOYSA-N | |
5-amino-3H-1,3,4-thiadiazole-2-thione | |
2723847 | |
98% | |
5-Amino-1, 3, 4-thiadiazole-2-thiol, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 219-078-4
missing translation for 'rtecsNumber' : XI4550000
missing translation for 'tsca' : TSCA
- 5-Amino-1,3,4-thiadiazole-2-thiol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.