missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific™

Product Code. 10232392
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51516-67-7
Molecular Formula C10H7ClN4
Molecular Weight (g/mol) 218.64
MDL Number MFCD00052031
InChI Key WIWSALMJHPGFDY-UHFFFAOYSA-N
Synonym 5-amino-1-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl, maybridge1_001852, acmc-1b1ld, 5-amino-1-4-chloro-phenyl-1h-pyrazole-4-carbonit, 1-4-chlorophenyl-5-amino-1h-pyrazole-4-carbonitrile
PubChem CID 639097
SMILES NC1=C(C=NN1C1=CC=C(Cl)C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
IUPAC Name 5-amino-1-(4-chlorophenyl)pyrazole-4-carbonitrile
Formula Weight 218.65
Percent Purity 97%
Chemical Name or Material 5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.