missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1-pentanol, 50 wt.% aqueous solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
800 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 48.0 |
CAS Max % | 52.0 |
Density | 0.9870g/mL |
Flash Point | >100°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | OH(CH2)5NH2 |
Quantity | 25 mL |
Specific Gravity | 0.987 |
Solubility Information | Solubility in water: miscible. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468947 | Certificate of Analysis | 31/01/2025 | 387910250,387911000,387918000 |
A0430435 | Certificate of Analysis | 18/11/2022 | 387910250,387911000,387918000 |
A0431125 | Certificate of Analysis | 14/06/2021 | 387910250,387911000,387918000 |
A0391474 | Certificate of Analysis | 30/11/2020 | 387910250,387911000,387918000 |
Safety and Handling


Product Identifier
- 5-Amino-1-pentanol, 50 wt.% aqueous solution
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Amino-1-pentanol, 50 wt.% aqueous solution, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction