missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-tetrazole monohydrate, 99%, Thermo Scientific Chemicals

Product Code. 11436936
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.

Solubility
Soluble in water, ethanol (1.36%).

Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15454-54-3
Molecular Formula CH5N5O
Molecular Weight (g/mol) 103.085
MDL Number MFCD00149327
InChI Key JVSMPWHQUPKRNV-UHFFFAOYSA-N
Synonym 1h-tetrazol-5-amine hydrate, 5-amino-1h-tetrazole monohydrate, 5-aminotetrazole monohydrate, 5-amino-1h-tetrazole hydrate, 5-aminotetrazole hydrate, 1h-1,2,3,4-tetrazol-5-amine hydrate, 2h-1,2,3,4-tetrazol-5-amine hydrate, tetrazol-5-amine, 5-aminotetrazole.h2o, 5-aminotetrazole h2o
PubChem CID 12211273
IUPAC Name 2H-tetrazol-5-amine;hydrate
SMILES C1(=NNN=N1)N.O

Specifications

Melting Point ∼203°C (decomposition)
pH 3.7
Quantity 500 g
UN Number UN1325
Beilstein 108872
Solubility Information Soluble in water,ethanol (1.36%).
Formula Weight 103.08 (85.06 Anhydrous)
Percent Purity 99%
Chemical Name or Material 5-Amino-1H-tetrazole monohydrate
compliance-icons compliance-icons
Product Identifier
  • 5-Amino-1H-tetrazole monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2H-Tetrazol-5-amine,hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.