missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4-difluorobenzoic acid, 98%, Thermo Scientific™

Product Code. 15404497
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 639858-45-0
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.119
MDL Number MFCD07368848
InChI Key AMOPVOVLPZDNMD-UHFFFAOYSA-N
Synonym 2,4-difluoro-5-aminobenzoic acid, 5-amino-2,4-difluoro-benzoic acid, benzoic acid, 5-amino-2,4-difluoro
PubChem CID 21049431
IUPAC Name 5-amino-2,4-difluorobenzoic acid
SMILES C1=C(C(=CC(=C1N)F)F)C(=O)O

Specifications

Melting Point ∽173°C (decomposition)
Quantity 5 g
Formula Weight 173.12
Percent Purity 98%
Chemical Name or Material 5-Amino-2,4-difluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'