Learn More
5-Amino-2-bromobenzoic acid, 95%, Thermo Scientific Chemicals
£165.00
Chemical Identifiers
CAS | 2840-02-0 |
---|---|
Molecular Formula | C7H6BrNO2 |
Molecular Weight (g/mol) | 216.03 |
MDL Number | MFCD07368968 |
InChI Key | FEXDUVBQBNYSQV-UHFFFAOYSA-N |
Synonym | 5-amino-2-bromo-benzoic acid, 4-bromo-3-carboxyaniline, benzoic acid,5-amino-2-bromo, benzoic acid, 5-amino-2-bromo, pubchem4637, acmc-209h2e, 5-amino-2-bromobenzoicacid, 2-bromo-5-aminobenzoic acid, ksc561g5h, 5-amino-2-bromo benzoic acid |
PubChem CID | 2769624 |
IUPAC Name | 5-amino-2-bromobenzoic acid |
SMILES | NC1=CC(C(O)=O)=C(Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426848
|
Thermo Scientific Alfa Aesar
H60382.14 |
25 g | N/A | ||||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
2840-02-0 | |
216.03 | |
FEXDUVBQBNYSQV-UHFFFAOYSA-N | |
2769624 | |
NC1=CC(C(O)=O)=C(Br)C=C1 |
C7H6BrNO2 | |
MFCD07368968 | |
5-amino-2-bromo-benzoic acid, 4-bromo-3-carboxyaniline, benzoic acid,5-amino-2-bromo, benzoic acid, 5-amino-2-bromo, pubchem4637, acmc-209h2e, 5-amino-2-bromobenzoicacid, 2-bromo-5-aminobenzoic acid, ksc561g5h, 5-amino-2-bromo benzoic acid | |
5-amino-2-bromobenzoic acid |
Specifications
2840-02-0 | |
C7H6BrNO2 | |
5-amino-2-bromo-benzoic acid, 4-bromo-3-carboxyaniline, benzoic acid,5-amino-2-bromo, benzoic acid, 5-amino-2-bromo, pubchem4637, acmc-209h2e, 5-amino-2-bromobenzoicacid, 2-bromo-5-aminobenzoic acid, ksc561g5h, 5-amino-2-bromo benzoic acid | |
FEXDUVBQBNYSQV-UHFFFAOYSA-N | |
5-amino-2-bromobenzoic acid | |
2769624 | |
95% |
171°C to 179°C | |
MFCD07368968 | |
Slightly soluble in water. | |
NC1=CC(C(O)=O)=C(Br)C=C1 | |
216.03 | |
216.03 | |
5-Amino-2-bromobenzoic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only