Learn More
5-Amino-2-chloro-4-methylpyridine, 98%, Thermo Scientific Chemicals
£48.10 - £184.00
Chemical Identifiers
CAS | 66909-38-4 |
---|---|
Molecular Formula | C6H7ClN2 |
Molecular Weight (g/mol) | 142.586 |
MDL Number | MFCD03427661 |
InChI Key | PLSMBLIGZJWLEJ-UHFFFAOYSA-N |
Synonym | 3-amino-6-chloro-4-picoline, 5-amino-2-chloro-4-methylpyridine, 5-amino-2-chloro-4-picoline, 3-amino-6-chloro-4-methyl-pyridine, 3-pyridinamine, 6-chloro-4-methyl, pubchem6206, 6-chloro-4-methyl-3-pyridinamine, 6-chloro-4-methyl-3-pyridylamine, 3-amino-6-chloro-4-methylpyridine, 6-chloro-4-methyl-3-pyridineamine |
PubChem CID | 2734425 |
IUPAC Name | 6-chloro-4-methylpyridin-3-amine |
SMILES | CC1=CC(=NC=C1N)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15427618
|
Thermo Scientific Alfa Aesar
H61115.06 |
5 g |
£48.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15437618
|
Thermo Scientific Alfa Aesar
H61115.14 |
25 g |
£184.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Amino-2-chloro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Amino-2-chloro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
66909-38-4 | |
142.586 | |
PLSMBLIGZJWLEJ-UHFFFAOYSA-N | |
2734425 | |
CC1=CC(=NC=C1N)Cl |
C6H7ClN2 | |
MFCD03427661 | |
3-amino-6-chloro-4-picoline, 5-amino-2-chloro-4-methylpyridine, 5-amino-2-chloro-4-picoline, 3-amino-6-chloro-4-methyl-pyridine, 3-pyridinamine, 6-chloro-4-methyl, pubchem6206, 6-chloro-4-methyl-3-pyridinamine, 6-chloro-4-methyl-3-pyridylamine, 3-amino-6-chloro-4-methylpyridine, 6-chloro-4-methyl-3-pyridineamine | |
6-chloro-4-methylpyridin-3-amine |
Specifications
66909-38-4 | |
C6H7ClN2 | |
3-amino-6-chloro-4-picoline, 5-amino-2-chloro-4-methylpyridine, 5-amino-2-chloro-4-picoline, 3-amino-6-chloro-4-methyl-pyridine, 3-pyridinamine, 6-chloro-4-methyl, pubchem6206, 6-chloro-4-methyl-3-pyridinamine, 6-chloro-4-methyl-3-pyridylamine, 3-amino-6-chloro-4-methylpyridine, 6-chloro-4-methyl-3-pyridineamine | |
PLSMBLIGZJWLEJ-UHFFFAOYSA-N | |
6-chloro-4-methylpyridin-3-amine | |
2734425 | |
98% | |
5-Amino-2-chloro-4-methylpyridine |
Orange | |
MFCD03427661 | |
Slightly soluble in water. | |
CC1=CC(=NC=C1N)Cl | |
142.586 | |
142.59 | |
Solid |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only