missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-fluorobenzoic acid, 99%, Thermo Scientific™

Product Code. 11336539
Click to view available options
Quantity:
500 mg
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56741-33-4
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00077449
InChI Key WYGAIOJWQDRBRJ-UHFFFAOYSA-N
Synonym 5-amino-2-fluorobenzioc acid, 3-amino-6-fluorobenzoic acid, 5-amino-2-fluoro-benzoic acid, buttpark 19\01-52, benzoic acid, 5-amino-2-fluoro, pubchem1325, 2-fluoro-5-aminobenzoic acid, ksc269g4f, 3-amino-6-fluoro-benzoic acid, rarechem al bo 1979
PubChem CID 2735910
IUPAC Name 5-amino-2-fluorobenzoic acid
SMILES NC1=CC(C(O)=O)=C(F)C=C1

Specifications

Melting Point 190°C to 192°C
Quantity 500 mg
Formula Weight 155.13
Percent Purity 99%
Chemical Name or Material 5-Amino-2-fluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'