missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 300°C |
Quantity | 250 mg |
Formula Weight | 185.42 |
Percent Purity | 95% |
Chemical Name or Material | 5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'