missing translation for 'onlineSavingsMsg'
Learn More
5-Amino-2-methyl-1,3-benzoxazole, 97%, Thermo Scientific Chemicals Product Code.: 10775652

5-Amino-2-methyl-1,3-benzoxazole, 97%, Thermo Scientific Chemicals

Product Code. 10775652
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10775652

Brand: Thermo Scientific Acros 437920010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72745-76-7
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.17
MDL Number MFCD00628970
InChI Key FECYTFWPNCBIHC-UHFFFAOYSA-N
Synonym 2-methylbenzo d oxazol-5-amine, 2-methyl-5-aminobenzoxazole, 5-amino-2-methylbenzoxazole, 5-benzoxazolamine,2-methyl, 2-methylbenzoxazole-5-ylamine, 2-methyl-benzoxazol-5-ylamine, 5-benzoxazolamine, 2-methyl, 2-methyl-benzooxazol-5-ylamine, 2-methylbenzoxazol-5-yl amine, 5-amino-2-methyl-1,3-benzoxazole
PubChem CID 737920
IUPAC Name 2-methyl-1,3-benzoxazol-5-amine
SMILES CC1=NC2=CC(N)=CC=C2O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 72°C to 74.6°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 148.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methyl-1, 3-benzoxazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345037 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number437920010,437920050
Lot Number336143 Certificate TypeCertificate of Analysis Date07/10/2020 Catalog Number437920010,437920050
compliance-icons
Product Identifier
  • 5-Amino-2-methyl-1,3-benzoxazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-2-methyl-1,3-benzoxazole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.