missing translation for 'onlineSavingsMsg'
Learn More
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals Product Code.: 15494167

5-Amino-3-(4-methoxyphenyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 15494167
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15494167

Brand: Thermo Scientific Alfa Aesar H31580.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19541-95-8
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.218
MDL Number MFCD00462192
InChI Key UPAGEJODHNVJNM-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-1h-pyrazol-5-amine, 5-4-methoxyphenyl-1h-pyrazol-3-amine, 3-amino-5-4-methoxyphenyl pyrazole, 5-amino-3-4-methoxyphenyl pyrazole, 3-amino-5-4-methoxyphenyl-1h-pyrazole, 5-4-methoxy-phenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-methoxyphenyl-1h-pyrazole, 1h-pyrazol-3-amine, 5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-pyrazol-3-amine, 5-4-methoxyphenyl-2h-pyrazol-3-ylamine
PubChem CID 2756469
IUPAC Name 5-(4-methoxyphenyl)-1H-pyrazol-3-amine
SMILES COC1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
Formula Weight 189.22
Percent Purity 97%
Chemical Name or Material 5-Amino-3-(4-methoxyphenyl)-1H-pyrazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10162620 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31580.03,H31580.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-3-(4-methoxyphenyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.