missing translation for 'onlineSavingsMsg'
Learn More

5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid, 97%, Thermo Scientific™

Product Code. 10406672
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10406672

Brand: Thermo Scientific Maybridge SB01487EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6453-58-3
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.312
MDL Number MFCD00063011
InChI Key RSYYQCDERUOEFI-UHFFFAOYSA-N
Synonym 5-amino imino methyl amino-2-benzoylamino pentanoic acid, 5-carbamimidamido-2-phenylformamido pentanoic acid, n2-benzoylarginine, l-arginine, n2-benzoyl, benzoyl-dl-arginine, n-?-benzoyl-l-arginine, 2-benzamido-5-guanidinopentanoic acid, 2-benzoylamino-5-guanidinovaleric acid, arginine,n2-benzoyl-,monohydrochloride 9ci, 2-benzamido-5-diaminomethylideneamino pentanoic acid
PubChem CID 273355
IUPAC Name 2-benzamido-5-(diaminomethylideneamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C6H5C(O)NHCH(CO2H)CH2CH2CH2NHC(NH)NH2
Quantity 10 g
Formula Weight 278.31
Percent Purity 97%
Chemical Name or Material 5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.