missing translation for 'onlineSavingsMsg'
Learn More

5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid, 97%, Thermo Scientific™

£8.10 - £55.20

Chemical Identifiers

CAS 6453-58-3
Molecular Formula C13H18N4O3
Molecular Weight (g/mol) 278.312
MDL Number MFCD00063011
InChI Key RSYYQCDERUOEFI-UHFFFAOYSA-N
Synonym 5-amino imino methyl amino-2-benzoylamino pentanoic acid, 5-carbamimidamido-2-phenylformamido pentanoic acid, n2-benzoylarginine, l-arginine, n2-benzoyl, benzoyl-dl-arginine, n-?-benzoyl-l-arginine, 2-benzamido-5-guanidinopentanoic acid, 2-benzoylamino-5-guanidinovaleric acid, arginine,n2-benzoyl-,monohydrochloride 9ci, 2-benzamido-5-diaminomethylideneamino pentanoic acid
PubChem CID 273355
IUPAC Name 2-benzamido-5-(diaminomethylideneamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10591312
View Documents Promotion Details
Thermo Scientific Maybridge
SB01487DA
1 g
£8.10
1g
Please to purchase this item. Need a web account? Register with us today!
10406672
View Documents Promotion Details
Thermo Scientific Maybridge
SB01487EA
10 g
£29.90
10g
Please to purchase this item. Need a web account? Register with us today!
11396925
View Documents Promotion Details
Thermo Scientific Maybridge
SB01487EB
25 g
£55.20
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6453-58-3
278.312
RSYYQCDERUOEFI-UHFFFAOYSA-N
273355
C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)O
C13H18N4O3
MFCD00063011
5-amino imino methyl amino-2-benzoylamino pentanoic acid, 5-carbamimidamido-2-phenylformamido pentanoic acid, n2-benzoylarginine, l-arginine, n2-benzoyl, benzoyl-dl-arginine, n-?-benzoyl-l-arginine, 2-benzamido-5-guanidinopentanoic acid, 2-benzoylamino-5-guanidinovaleric acid, arginine,n2-benzoyl-,monohydrochloride 9ci, 2-benzamido-5-diaminomethylideneamino pentanoic acid
2-benzamido-5-(diaminomethylideneamino)pentanoic acid

Specifications

6453-58-3
97%
C13H18N4O3
MFCD00063011
RSYYQCDERUOEFI-UHFFFAOYSA-N
2-benzamido-5-(diaminomethylideneamino)pentanoic acid
273355
97%
97.0
Amber glass bottle
C6H5C(O)NHCH(CO2H)CH2CH2CH2NHC(NH)NH2
5-amino imino methyl amino-2-benzoylamino pentanoic acid, 5-carbamimidamido-2-phenylformamido pentanoic acid, n2-benzoylarginine, l-arginine, n2-benzoyl, benzoyl-dl-arginine, n-?-benzoyl-l-arginine, 2-benzamido-5-guanidinopentanoic acid, 2-benzoylamino-5-guanidinovaleric acid, arginine,n2-benzoyl-,monohydrochloride 9ci, 2-benzamido-5-diaminomethylideneamino pentanoic acid
C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)O
278.312
278.31
5-[[Amino(imino)methyl]amino]-2-(benzoylamino)pentanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications