missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
5-Aminoisophthalic acid is a useful biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Aminoisophthalic acid is a useful biochemical for proteomics research.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
5-Aminoisophthalic acid is a useful biochemical for proteomics research.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Quantity | 250 g |
Beilstein | 2805628 |
Solubility Information | Insoluble in water. |
Formula Weight | 181.15 |
Percent Purity | 95% |
Chemical Name or Material | 5-Aminoisophthalic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'