missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1H-benzimidazole, 97%, Thermo Scientific Chemicals
£99.80 - £342.00
Chemical Identifiers
CAS | 4887-88-1 |
---|---|
Molecular Formula | C7H5BrN2 |
Molecular Weight (g/mol) | 197.04 |
MDL Number | MFCD00160001 |
InChI Key | GEDVWGDBMPJNEV-UHFFFAOYSA-N |
Synonym | 5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618 |
PubChem CID | 785299 |
SMILES | BrC1=CC=C2N=CNC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15254884
|
Thermo Scientific Acros
451670050 |
5 g |
£99.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15274884
|
Thermo Scientific Acros
451670250 |
25 g |
£342.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4887-88-1 | |
197.04 | |
GEDVWGDBMPJNEV-UHFFFAOYSA-N | |
785299 |
C7H5BrN2 | |
MFCD00160001 | |
5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618 | |
BrC1=CC=C2N=CNC2=C1 |
Specifications
4887-88-1 | |
96% min. (HPLC) | |
C7H5BrN2 | |
5-bromo-1h-benzimidazole, 5-bromo-1h-benzo d imidazole, 5-bromo-1h-1,3-benzodiazole, 5-bromobenzimidazole, 1h-benzimidazole, 5-bromo, 5-bromo-3h-1,3-benzodiazole, 6-bromobenzimidazole, 5-bromo-benzimidazole, 6-bromo benzimidazole, pubchem9618 | |
BrC1=CC=C2N=CNC2=C1 | |
197.04 | |
197.03 | |
5-Bromo-1H-benzimidazole |
130°C to 134°C | |
Glass bottle | |
MFCD00160001 | |
GEDVWGDBMPJNEV-UHFFFAOYSA-N | |
6-bromo-1H-benzimidazole | |
785299 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid contact during pregnancy/while nursing.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Bromo-1H-benzimidazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only