Learn More
5-Bromo-2,1,3-benzothiadiazole, 97%, Thermo Scientific™
£38.00 - £148.00
Chemical Identifiers
CAS | 1753-75-9 |
---|---|
Molecular Formula | C6H3BrN2S |
Molecular Weight (g/mol) | 215.068 |
MDL Number | MFCD00460091 |
InChI Key | LLCRUZDFDGTAAN-UHFFFAOYSA-N |
Synonym | 5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole |
PubChem CID | 2776295 |
IUPAC Name | 5-bromo-2,1,3-benzothiadiazole |
SMILES | C1=CC2=NSN=C2C=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10652053
|
Thermo Scientific Maybridge
CC09210DA |
1 g |
£38.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10473084
|
Thermo Scientific Maybridge
CC09210DE |
5 g |
£148.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
1753-75-9 | |
215.068 | |
LLCRUZDFDGTAAN-UHFFFAOYSA-N | |
2776295 | |
C1=CC2=NSN=C2C=C1Br |
C6H3BrN2S | |
MFCD00460091 | |
5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole | |
5-bromo-2,1,3-benzothiadiazole |
Specifications
1753-75-9 | |
100.0 | |
Amber glass bottle | |
MFCD00460091 | |
LLCRUZDFDGTAAN-UHFFFAOYSA-N | |
5-bromo-2,1,3-benzothiadiazole | |
2776295 | |
97% |
96.5 | |
97% | |
C6H3BrN2S | |
5-bromobenzo c 1,2,5 thiadiazole, 5-bromobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole,5-bromo, 5-bromobenzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole, 5-bromo, pubchem12684, acmc-209ea1, amth068, 5-bromobenz-2,1,3-thiadiazole, 5-bromo-benzo 1,2,5 thiadiazole | |
C1=CC2=NSN=C2C=C1Br | |
215.068 | |
215.07 | |
5-Bromo-2, 1, 3-benzothiadiazole |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- 5-Bromo-2,1,3-benzothiadiazole
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.