missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2,3-dihydrobenzo[b]furan-7-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 227.06 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-2, 3-dihydrobenzo[b]furan-7-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'