Learn More
5-Bromo-2,3-dihydrobenzo[b]furan-7-carboxylic acid, 97%, Thermo Scientific™
£27.60 - £109.00
Chemical Identifiers
CAS | 41177-72-4 |
---|---|
Molecular Formula | C9H7BrO3 |
Molecular Weight (g/mol) | 243.06 |
MDL Number | MFCD00191391 |
InChI Key | LEBMKAXASFPSFA-UHFFFAOYSA-N |
Synonym | 5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid |
PubChem CID | 2794797 |
IUPAC Name | 5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid |
SMILES | OC(=O)C1=CC(Br)=CC2=C1OCC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10432242
|
Thermo Scientific Maybridge
MO07139DA |
1 g | Amber Glass Bottle |
£27.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10718065
|
Thermo Scientific Maybridge
MO07139DE |
5 g | Amber Glass Bottle |
£109.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
41177-72-4 | |
243.06 | |
LEBMKAXASFPSFA-UHFFFAOYSA-N | |
2794797 | |
OC(=O)C1=CC(Br)=CC2=C1OCC2 |
C9H7BrO3 | |
MFCD00191391 | |
5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid | |
5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid |
Specifications
41177-72-4 | |
97% | |
MFCD00191391 | |
LEBMKAXASFPSFA-UHFFFAOYSA-N | |
5-bromo-2,3-dihydro-1-benzofuran-7-carboxylic acid | |
2794797 | |
97% |
97.0 | |
C9H7BrO3 | |
5-bromo-2,3-dihydrobenzofuran-7-carboxylic acid, 5-bromo-2,3-dihydrobenzo b furan-7-carboxylic acid, 5-bromocoumaran-7-carboxylic acid, 5-bromo-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylic acid, 5-bromo-2,3-dihydro, zlchem 1161, acmc-1aqu8, 5-bromo-2,3-dihydrobenzofuran-7-carboxylicacid, 5-bromanyl-2,3-dihydro-1-benzofuran-7-carboxylic acid | |
OC(=O)C1=CC(Br)=CC2=C1OCC2 | |
243.06 | |
243.06 | |
5-Bromo-2, 3-dihydrobenzo[b]furan-7-carboxylic acid, May contain varying amounts of anhydride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.