missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dimethoxybenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15467078
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32246-20-1
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.071
MDL Number MFCD05865172
InChI Key WOPJFSQYDOHZMK-UHFFFAOYSA-N
Synonym 5-bromo-2 4-dimethoxybenzoic acid, acmc-20am5v, 5-bromo-2,4-dimethoxybenzoicacid, 2,4-dimethoxy-5-bromobenzoic acid, 2,4-dimethoxy-5-bromo-benzoic acid, 5-bromo-2,4-dimethoxy-benzoic acid, 5-bromanyl-2,4-dimethoxy-benzoic acid, benzoic acid, 5-bromo-2,4-dimethoxy, 5-bromo-2,4-dimethoxybenzoic acid
PubChem CID 13505097
IUPAC Name 5-bromo-2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1C(=O)O)Br)OC

Specifications

Melting Point 198°C to 202°C
Quantity 5 g
Formula Weight 261.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-2,4-dimethoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'