missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloropyrimidine, 96%, Thermo Scientific Chemicals

Product Code. 10189662
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32779-36-5
Molecular Formula C4H2BrClN2
Molecular Weight (g/mol) 193.43
MDL Number MFCD00483232
InChI Key XPGIBDJXEVAVTO-UHFFFAOYSA-N
Synonym 2-chloro-5-bromopyrimidine, 5-bromo-2-chloro-pyrimidine, 5-bromo-2-chloropyrimidne, pyrimidine, 5-bromo-2-chloro, 5-bromo-2-chloro-1,3-diazine, pubchem5159, zlchem 1210, acmc-209hvm, 5-bromo chloropyrimidine, 5-bromo-2-chloroprimidine
PubChem CID 606665
IUPAC Name 5-bromo-2-chloropyrimidine
SMILES ClC1=NC=C(Br)C=N1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 78.0°C to 80.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble in water
Formula Weight 193.43
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-chloropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'