missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-deoxycytidine, 99%

Product Code. 15446909
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15446909

Brand: Thermo Scientific Alfa Aesar J65456.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1022-79-3 | C9H12BrN3O4 | 306.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1022-79-3
Color White to Yellow
Molecular Formula C9H12BrN3O4
MDL Number MFCD00047496
Quantity 1 g
Synonym 5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy
InChI Key KISUPFXQEHWGAR-RRKCRQDMSA-N
SMILES C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O
IUPAC Name 4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Molecular Weight (g/mol) 306.116
PubChem CID 472175
Formula Weight 307.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Bromo-2'-deoxycytidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.