missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-hydroxy-3-methoxybenzyl alcohol

£46.01 - £138.00

Chemical Identifiers

CAS 35090-64-3
Molecular Formula C8H9BrO3
Molecular Weight (g/mol) 233.06
MDL Number MFCD02081986
InChI Key UBLQIISPEAEAIX-UHFFFAOYSA-N
Synonym 4-bromo-2-hydroxymethyl-6-methoxyphenol, 4-bromo-2-hydroxymethyl-6-methoxybenzenol, acmc-20amo4, benzenemethanol,5-bromo-2-hydroxy-3-methoxy
PubChem CID 894092
IUPAC Name 4-bromo-2-(hydroxymethyl)-6-methoxyphenol
SMILES COC1=C(O)C(CO)=CC(Br)=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430040
View Documents Promotion Details
Alfa Aesar™
H50569.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11440040
View Documents Promotion Details
Alfa Aesar™
H50569.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

35090-64-3
233.06
UBLQIISPEAEAIX-UHFFFAOYSA-N
894092
COC1=C(O)C(CO)=CC(Br)=C1
C8H9BrO3
MFCD02081986
4-bromo-2-hydroxymethyl-6-methoxyphenol, 4-bromo-2-hydroxymethyl-6-methoxybenzenol, acmc-20amo4, benzenemethanol,5-bromo-2-hydroxy-3-methoxy
4-bromo-2-(hydroxymethyl)-6-methoxyphenol

Specifications

35090-64-3
MFCD02081986
UBLQIISPEAEAIX-UHFFFAOYSA-N
4-bromo-2-(hydroxymethyl)-6-methoxyphenol
894092
5-Bromo-2-hydroxy-3-methoxybenzyl alcohol
C8H9BrO3
4-bromo-2-hydroxymethyl-6-methoxyphenol, 4-bromo-2-hydroxymethyl-6-methoxybenzenol, acmc-20amo4, benzenemethanol,5-bromo-2-hydroxy-3-methoxy
COC1=C(O)C(CO)=CC(Br)=C1
233.06
233.06
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications