missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodotoluene, 98+%, Thermo Scientific Chemicals

Product Code. 11359739
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Bromo-2-iodotoluene is used as a medical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-iodotoluene is used as a medical intermediate and in organic synthesis.

Notes
Light sensitive. Incompatible with Strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116632-39-4
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
MDL Number MFCD00060664
InChI Key GHTUADBHTFHMNI-UHFFFAOYSA-N
Synonym 5-bromo-2-iodotoluene, 1-bromo-4-iodo-5-methylbenzene, benzene, 4-bromo-1-iodo-2-methyl, bromoiodotoluene, pubchem3793, 2-iodo-5-bromo-toluene, 5-bromo-2-iodo toluene, 5-bromo-2-iodotoluene,, 4-bromo-2-methyliodobenzene, 4-bromo-2-methyl-iodobenzene
PubChem CID 2724601
IUPAC Name 4-bromo-1-iodo-2-methylbenzene
SMILES CC1=CC(Br)=CC=C1I

Specifications

Density 2.139
Boiling Point 262°C to 265°C
Flash Point >110°C (230°F)
Refractive Index 1.6515
Quantity 5 g
Beilstein 2325157
Sensitivity Light sensitive
Formula Weight 296.93
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-2-iodotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'