missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxyphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10351100
5 g, Glass bottle
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 89694-45-1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 136.4°C to 137.4°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H8BBrO3
MDL Number MFCD00093409
Quantity 5 g
Synonym 5-bromo-2-methoxyphenyl boronic acid, 5-bromo-2-methoxybenzeneboronic acid, 4-bromoanisole-2-boronic acid, 3-bromo-6-methoxyphenylboronic acid, boronic acid,b-5-bromo-2-methoxyphenyl, 5-bromo-2-methoxyphenylboronicacid, rarechem ah pb 0045, acmc-209r2g, amtb789, 2-methoxy-5-bromophenylboronic acid
InChI Key FVRLSHRAYPFXRQ-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)Br)OC)(O)O
IUPAC Name (5-bromo-2-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 230.85
PubChem CID 4532730
Formula Weight 230.85
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-methoxyphenylboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-2-methoxyphenylboronic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.