missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methoxyquinoline, 96%, Thermo Scientific Chemicals

POA - £620.00

Chemical Identifiers

CAS 776296-12-9
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.084
MDL Number MFCD16038652
InChI Key YNVIAXULFXXTLV-UHFFFAOYSA-N
Synonym 5-bromo-3-methoxy-quinoline, 5-bromo-3-methyoxy-quinoline, quinoline, 5-bromo-3-methoxy
PubChem CID 58227851
IUPAC Name 5-bromo-3-methoxyquinoline
SMILES COC1=CN=C2C=CC=C(C2=C1)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15411068
View Documents
Thermo Scientific Alfa Aesar
H54919.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15491058
View Documents
Thermo Scientific Alfa Aesar
H54919.03
1 g
£157.00
1g
Please to purchase this item. Need a web account? Register with us today!
15401068
View Documents
Thermo Scientific Alfa Aesar
H54919.06
5 g
£620.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Notes
Refrigerated (2-4°C)
Specifications

Chemical Identifiers

776296-12-9
238.084
YNVIAXULFXXTLV-UHFFFAOYSA-N
58227851
COC1=CN=C2C=CC=C(C2=C1)Br
C10H8BrNO
MFCD16038652
5-bromo-3-methoxy-quinoline, 5-bromo-3-methyoxy-quinoline, quinoline, 5-bromo-3-methoxy
5-bromo-3-methoxyquinoline

Specifications

776296-12-9
MFCD16038652
YNVIAXULFXXTLV-UHFFFAOYSA-N
5-bromo-3-methoxyquinoline
58227851
96%
C10H8BrNO
5-bromo-3-methoxy-quinoline, 5-bromo-3-methyoxy-quinoline, quinoline, 5-bromo-3-methoxy
COC1=CN=C2C=CC=C(C2=C1)Br
238.084
238.08
5-Bromo-3-methoxyquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only