missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine, 95%, Thermo Scientific™

Product Code. 11352399
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446299-84-9
Molecular Formula C11H15BrN2O
Molecular Weight (g/mol) 271.158
MDL Number MFCD09037447
InChI Key DJAJPKVIGCOGCY-UHFFFAOYSA-N
Synonym n-5-bromo-3-methylpyridin-2-yl pivalamide, 5-bromo-2-pivaloylamino-3-picoline, n-5-bromo-3-methylpyridin-2-yl-2,2-dimethylpropanamide, 5-bromo-3-methyl-2-2,2,2-trimethylacetamido pyridine, n-5-bromo-3-methyl-2-pyridyl pivalamide, 5-bromo-3-methyl-2-pivaloylamino pyridine, n-5-bromo-3-methyl-2-pyridinyl-2,2-dimethylpropanamide
PubChem CID 42553022
IUPAC Name N-(5-bromo-3-methylpyridin-2-yl)-2,2-dimethylpropanamide
SMILES CC1=CC(=CN=C1NC(=O)C(C)(C)C)Br

Specifications

Melting Point 125°C to 128°C
Quantity 1 g
Formula Weight 271.16
Percent Purity 95%
Chemical Name or Material 5-Bromo-3-methyl-2-(2,2,2-trimethylacetamido)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'