missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-pyridineacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11303157
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39891-12-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00829308
InChI Key UETIDNDXXGCJCE-UHFFFAOYSA-N
Synonym 5-bromo-3-pyridylacetic acid, 2-5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridineacetic acid, 3-pyridineacetic acid, 5-bromo, 2-5-bromo-3-pyridyl acetic acid, 5-bromo-3-pyridylaceticacid, 5-bromo-pyridin-3-yl-acetic acid, 5-bromopyridin-3-yl acetic acid, 5-bromo-3-pyridinyl acetic acid, 2-5-bromopyridin-3-yl acetic acid
PubChem CID 2802539
IUPAC Name 2-(5-bromopyridin-3-yl)acetic acid
SMILES C1=C(C=NC=C1Br)CC(=O)O

Specifications

Melting Point 182°C to 185°C
Quantity 5 g
Beilstein 388264
Solubility Information Soluble in water.
Formula Weight 216.03
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-3-pyridineacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'