missing translation for 'onlineSavingsMsg'
Learn More
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals Product Code.: 15406909

5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals

Product Code. 15406909
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15406909

Brand: Thermo Scientific Alfa Aesar J65443.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Bromo-5-nitro-1,3-dioxane, also known as bronidox, shows antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi.

5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth

Application

• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30007-47-7
Molecular Formula C4H6BrNO4
Molecular Weight (g/mol) 211.999
MDL Number MFCD00101855
InChI Key XVBRCOKDZVQYAY-UHFFFAOYSA-N
Synonym bronidox, 1,3-dioxane, 5-bromo-5-nitro, 5-bromo-5-nitro-m-dioxane, unii-u184i9qbnm, 5-brom-5-nitro-1,3-dioxan, m-dioxane, 5-bromo-5-nitro, 5-brom-5-nitro-1,3-dioxan german, u184i9qbnm, bronidox l, acmc-209hcf
PubChem CID 1807
IUPAC Name 5-bromo-5-nitro-1,3-dioxane
SMILES C1C(COCO1)([N+](=O)[O-])Br

Specifications

Melting Point 58°C to 61°C
Color White
Quantity 5 g
Solubility Information Soluble in water at 12.5mg/ml
Formula Weight 212
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-5-nitro-1,3-dioxane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP10L002 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberJ65443.14,J65443.06
Lot NumberM28L052 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberJ65443.14,J65443.06
Lot NumberZ24K034 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberJ65443.14,J65443.06
Lot Number10252980 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog NumberJ65443.14,J65443.06
Lot NumberX09K052 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberJ65443.14,J65443.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.