missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-6-methylpyridin-2-amine, 97%, Thermo Scientific™

£43.70 - £242.00

Chemical Identifiers

CAS 42753-71-9
Molecular Formula C6H8BrN2
Molecular Weight (g/mol) 188.05
MDL Number MFCD00068230
InChI Key SEOZHXRTVJPQPZ-UHFFFAOYSA-O
Synonym 2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine
PubChem CID 170678
IUPAC Name 5-bromo-6-methylpyridin-2-amine
SMILES CC1=[NH+]C(N)=CC=C1Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10102994
View Documents
Thermo Scientific Maybridge
XBX00110EA
10 g
£43.70
10g
Please to purchase this item. Need a web account? Register with us today!
10448604
View Documents
Thermo Scientific Maybridge
XBX00110EB
25 g
£79.40
25g
Please to purchase this item. Need a web account? Register with us today!
10225434
View Documents
Thermo Scientific Maybridge
XBX00110FA
100 g
£242.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

42753-71-9
188.05
SEOZHXRTVJPQPZ-UHFFFAOYSA-O
170678
CC1=[NH+]C(N)=CC=C1Br
C6H8BrN2
MFCD00068230
2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine
5-bromo-6-methylpyridin-2-amine

Specifications

42753-71-9
95%
C6H8BrN2
2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine
CC1=[NH+]C(N)=CC=C1Br
188.05
187.04
5-Bromo-6-methylpyridin-2-amine
95.0
Amber glass bottle
MFCD00068230
SEOZHXRTVJPQPZ-UHFFFAOYSA-O
5-bromo-6-methylpyridin-2-amine
170678
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications