missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromobenzo[b]furan-2-carbonyl chloride, 97%, Thermo Scientific™
£29.95 - £247.00
Chemical Identifiers
CAS | 62878-96-0 |
---|---|
Molecular Formula | C9H4BrClO2 |
Molecular Weight (g/mol) | 259.483 |
InChI Key | FGPAEHRRVKLUHA-UHFFFAOYSA-N |
Synonym | 5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride |
PubChem CID | 2795534 |
IUPAC Name | 5-bromo-1-benzofuran-2-carbonyl chloride |
SMILES | C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10045762
|
Thermo Scientific Maybridge
CC31202CB |
250 mg |
£29.95
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10219282
|
Thermo Scientific Maybridge
CC31202DA |
1 g |
£81.75
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10123724
|
Thermo Scientific Maybridge
CC31202DE |
5 g |
£247.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
62878-96-0 | |
259.483 | |
5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride | |
5-bromo-1-benzofuran-2-carbonyl chloride |
C9H4BrClO2 | |
FGPAEHRRVKLUHA-UHFFFAOYSA-N | |
2795534 | |
C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl |
Specifications
62878-96-0 | |
Amber Glass Bottle | |
5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride | |
C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl | |
259.483 | |
259.49 | |
5-Bromobenzo[b]furan-2-carbonyl chloride |
95% | |
C9H4BrClO2 | |
FGPAEHRRVKLUHA-UHFFFAOYSA-N | |
5-bromo-1-benzofuran-2-carbonyl chloride | |
2795534 | |
95% |