missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromobenzo[b]furan-2-carboxylic acid 97+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 241.04 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-1-benzofuran-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'