missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromobenzo[b]thiophene-2-carboxylic acid, ≥97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 257.11 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-1-benzothiophene-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'