missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-3-acetic acid, 99%, Thermo Scientific Chemicals

Product Code. 10244370
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40432-84-6
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.08
MDL Number MFCD00005637
InChI Key WTFGHMZUJMRWBK-UHFFFAOYSA-N
Synonym 5-bromoindole-3-acetic acid, 2-5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 1h-indole-3-acetic acid, 5-bromo, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-3-indoleacetic acid, chembl82440, 5-bromo-1h-indole-3-acetic acid, 5-bromoindole-3-aceticacid, 4ojq
PubChem CID 96734
IUPAC Name 2-(5-bromo-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 141°C to 148°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 254.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-3-acetic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'