missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoindole-3-acetic acid, 99%, Thermo Scientific Chemicals
£39.50 - £104.00
Chemical Identifiers
CAS | 40432-84-6 |
---|---|
Molecular Formula | C10H8BrNO2 |
Molecular Weight (g/mol) | 254.08 |
MDL Number | MFCD00005637 |
InChI Key | WTFGHMZUJMRWBK-UHFFFAOYSA-N |
Synonym | 5-bromoindole-3-acetic acid, 2-5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 1h-indole-3-acetic acid, 5-bromo, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-3-indoleacetic acid, chembl82440, 5-bromo-1h-indole-3-acetic acid, 5-bromoindole-3-aceticacid, 4ojq |
PubChem CID | 96734 |
IUPAC Name | 2-(5-bromo-1H-indol-3-yl)acetic acid |
SMILES | C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10589980
|
Thermo Scientific Acros
107072500 |
250 mg | Glass bottle | ||||||
10244370
|
Thermo Scientific Acros
107070010 |
1 g | Glass bottle |
£104.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40432-84-6 | |
254.08 | |
WTFGHMZUJMRWBK-UHFFFAOYSA-N | |
96734 | |
C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O |
C10H8BrNO2 | |
MFCD00005637 | |
5-bromoindole-3-acetic acid, 2-5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 1h-indole-3-acetic acid, 5-bromo, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-3-indoleacetic acid, chembl82440, 5-bromo-1h-indole-3-acetic acid, 5-bromoindole-3-aceticacid, 4ojq | |
2-(5-bromo-1H-indol-3-yl)acetic acid |
Specifications
40432-84-6 | |
100.0 | |
White to Beige | |
99% | |
MFCD00005637 | |
Solubility in water: insoluble in water | |
C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O | |
254.08 | |
254.08 | |
Crystalline Powder |
98.5 | |
141°C to 148°C | |
Authentic | |
C10H8BrNO2 | |
5-bromoindole-3-acetic acid, 2-5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 1h-indole-3-acetic acid, 5-bromo, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-3-indoleacetic acid, chembl82440, 5-bromo-1h-indole-3-acetic acid, 5-bromoindole-3-aceticacid, 4ojq | |
WTFGHMZUJMRWBK-UHFFFAOYSA-N | |
2-(5-bromo-1H-indol-3-yl)acetic acid | |
96734 | |
99% | |
5-Bromoindole-3-acetic acid, 99% |
Safety and Handling
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 254-917-8
RUO – Research Use Only