missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoindole, 99%, Thermo Scientific Chemicals
£18.39 - £130.65
Chemical Identifiers
CAS | 10075-50-0 |
---|---|
Molecular Formula | C8H6BrN |
Molecular Weight (g/mol) | 196.05 |
MDL Number | MFCD00005670 |
InChI Key | VXWVFZFZYXOBTA-UHFFFAOYSA-N |
Synonym | 5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661 |
PubChem CID | 24905 |
IUPAC Name | 5-bromo-1H-indole |
SMILES | BrC1=CC=C2NC=CC2=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10080640
|
Thermo Scientific Acros
107060050 |
5 g | Glass bottle |
£18.39
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10466900
|
Thermo Scientific Acros
107060250 |
25 g | Glass bottle |
£40.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10504284
|
Thermo Scientific Acros
107061000 |
100 g | Glass bottle |
£130.65
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10075-50-0 | |
196.05 | |
VXWVFZFZYXOBTA-UHFFFAOYSA-N | |
24905 | |
BrC1=CC=C2NC=CC2=C1 |
C8H6BrN | |
MFCD00005670 | |
5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661 | |
5-bromo-1H-indole |
Specifications
10075-50-0 | |
100.0 | |
White to Brown | |
98.5% min. (GC) | |
MFCD00005670 | |
VXWVFZFZYXOBTA-UHFFFAOYSA-N | |
5-bromo-1H-indole | |
24905 | |
99% | |
5-Bromoindole, 99% |
98.5 | |
89.0°C to 92.0°C | |
Authentic | |
C8H6BrN | |
5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661 | |
BrC1=CC=C2NC=CC2=C1 | |
196.05 | |
196.05 | |
Chunks and/or Powder |
Safety and Handling
missing translation for 'einecsNumber' : 233-208-7
Product Identifier
- 5-Bromoindole
RUO – Research Use Only