missing translation for 'onlineSavingsMsg'
Learn More

5-Bromonicotinamide, 98%, Thermo Scientific™

Product Code. 11433438
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28733-43-9
Molecular Formula C6H5BrN2O
Molecular Weight (g/mol) 201.023
MDL Number MFCD00173919
InChI Key YOQRXZIMSKLRCY-UHFFFAOYSA-N
Synonym 5-bromonicotinamide, 5-bromo-3-pyridinecarboxamide, nicotinamide, 5-bromo, 3-pyridinecarboxamide, 5-bromo, 5-bromo-nicotinamide, brt, pubchem1212, acmc-1cfs0, 3-bromo-5-carbamoylpyridine, 5-22-02-00182 beilstein handbook reference
PubChem CID 1808
IUPAC Name 5-bromopyridine-3-carboxamide
SMILES C1=C(C=NC=C1Br)C(=O)N

Specifications

Melting Point 226°C to 227°C
Quantity 25 g
Formula Weight 201.03
Percent Purity 98%
Chemical Name or Material 5-Bromonicotinamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.