missing translation for 'onlineSavingsMsg'
Learn More

(5-Bromopyrid-2-yl)methanol, 97%, Thermo Scientific™

£17.30 - £140.00

Chemical Identifiers

CAS 88139-91-7
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04035597
InChI Key RUCZFWMEACWFER-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxymethylpyridine, 2-hydroxymethyl-5-bromopyridine, 5-bromopyridin-2-yl methanol, 5-bromo-2-hydroxymethyl pyridine, 5-bromopyrid-2-yl methanol, 5-bromo-2-pyridinemethanol, 5-bromo-pyridin-2-yl-methanol, 2-pyridinemethanol, 5-bromo, 5-bromo-2-pyridyl methan-1-ol, 5-bromopyridine-2-methanol
PubChem CID 5200169
IUPAC Name (5-bromopyridin-2-yl)methanol
SMILES OCC1=CC=C(Br)C=N1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10577854
SDS View Documents
Thermo Scientific Maybridge
MO07897CB
250 mg
£17.30
250mg
Please to purchase this item. Need a web account? Register with us today!
10235414
SDS View Documents
Thermo Scientific Maybridge
MO07897DA
1 g
£65.60
1g
Please to purchase this item. Need a web account? Register with us today!
10113084
SDS View Documents
Thermo Scientific Maybridge
MO07897DE
5 g
£140.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(5-Bromopyrid-2-yl)methanol, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.