missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-sulfonyl chloride, 96%, Thermo Scientific Chemicals

Product Code. 11487656
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55854-46-1
Molecular Formula C4H2BrClO2S2
Molecular Weight (g/mol) 261.53
MDL Number MFCD00084909
InChI Key WGYBIEOLAFYDEC-UHFFFAOYSA-N
Synonym 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride
PubChem CID 2733924
IUPAC Name 5-bromothiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(Br)S1

Specifications

Melting Point 39°C to 42°C
Boiling Point 99°C to 101°C (0.5 mmHg)
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 261.55
Percent Purity 98%
Chemical Name or Material 5-Bromothiophene-2-sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.