Learn More
5-Bromovaleryl chloride, 98%, Thermo Scientific Chemicals
£48.40 - £153.00
Chemical Identifiers
CAS | 4509-90-4 |
---|---|
Molecular Formula | C14H11N3O2 |
Molecular Weight (g/mol) | 253.26 |
MDL Number | MFCD00013660 |
InChI Key | QANQQXSQVQYQFI-UHFFFAOYSA-N |
Synonym | 5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride |
PubChem CID | 521173 |
IUPAC Name | 5-bromopentanoyl chloride |
SMILES | CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449186
|
Thermo Scientific Alfa Aesar
A18046.06 |
5 g |
£48.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459186
|
Thermo Scientific Alfa Aesar
A18046.14 |
25 g |
£153.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
Chemical Identifiers
4509-90-4 | |
253.26 | |
QANQQXSQVQYQFI-UHFFFAOYSA-N | |
521173 | |
CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1 |
C14H11N3O2 | |
MFCD00013660 | |
5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride | |
5-bromopentanoyl chloride |
Specifications
4509-90-4 | |
43°C to 45°C (0.2 mmHg) | |
Irritating | |
Br(CH2)4COCl | |
MFCD00013660 | |
1745663 | |
5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride | |
QANQQXSQVQYQFI-UHFFFAOYSA-N | |
5-bromopentanoyl chloride | |
521173 | |
98% |
1.49 | |
107°C (225°F) | |
C14H11N3O2 | |
1.491 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1 | |
253.26 | |
199.46 | |
5-Bromovaleryl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only