missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-2702681
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10651673

Brand: Thermo Scientific Maybridge CD10088DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 88398-93-0
Molecular Formula C5H6Cl2N2O2S
Molecular Weight (g/mol) 229.075
MDL Number MFCD00052536
InChI Key HWGVUNSKAPCFNF-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole-4-sulfonyl chloride, 5-chloro-1,3-dimethyl-4-pyrazolesulfonyl chloride, 5-chloro-1,3-dimethyl-1h-pyrazole-4-sulphonyl chloride, 5-chloro-1,3-dimethyl-pyrazole-4-sulfonyl chloride, 5-chloro-1,3-dimethylpyrazole-4-sulfonylchloride, 5-chloranyl-1,3-dimethyl-pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride, 5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazol-4yl-sulfonyl chloride, 5-chloro-1,3-dimethyl-1h-pyrazol-4-yl sulfonyl chloride, 5-chloro-1,3-dimethyl-1h-pyrazol-4-ylsulphonyl chloride
PubChem CID 2777126
IUPAC Name 5-chloro-1,3-dimethylpyrazole-4-sulfonyl chloride
SMILES CC1=NN(C(=C1S(=O)(=O)Cl)Cl)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 229.09
Percent Purity 97%
Chemical Name or Material 5-chloro-1, 3-dimethyl-1H-pyrazole-4-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.