missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, Thermo Scientific™

Product Code. 10101072
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10101072

Brand: Thermo Scientific Maybridge GK02622DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 128455-63-0
Molecular Formula C6H4ClF3N2O2
Molecular Weight (g/mol) 228.555
MDL Number MFCD00215443
InChI Key IKGVBNQPAJOSFP-UHFFFAOYSA-N
Synonym 5-chloro-1-methyl-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 5-chloro-1-methyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 5-chloro-1-methyl-3-trifluoromethyl, pubchem16959, acmc-1c8gv, chloromethyltrifluoromethylpyrazolecarboxylicacid, 4-carboxy-5-chloro-1-methyl-3-trifluoromethyl-1h-pyrazole, 5-chloro-1-methyl-3-trifluoromethylpyrazol-4-carboxylic acid, 1h-pyrazole-4-carboxylic acid, 5-chloro-1-methyl-3-trifluoromethyl
PubChem CID 2773806
IUPAC Name 5-chloro-1-methyl-3-(trifluoromethyl)pyrazole-4-carboxylic acid
SMILES CN1C(=C(C(=N1)C(F)(F)F)C(=O)O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 228.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.