missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid, 98%

Product Code. 15410306
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 871329-55-4
Molecular Formula C9H10BClO4
Molecular Weight (g/mol) 228.44
MDL Number MFCD07363744
InChI Key SGPMCOBGNHZGJJ-UHFFFAOYSA-N
Synonym 5-chloro-2-ethoxycarbonyl phenylboronic acid, 5-chloro-2-ethoxycarbonyl phenyl boronic acid, 5-chloro-2-ethoxycarbonyl benzeneboronic acid, benzoic acid,2-borono-4-chloro-, 1-ethyl ester 9ci, ethyl 2-borono-4-chlorobenzoate, acmc-209qg0, 5-chloro-2-ethoxycarbonylphenyl boronic acid, 5-chloranyl-2-ethoxycarbonyl-phenyl boronic acid
PubChem CID 44784921
IUPAC Name (5-chloro-2-ethoxycarbonylphenyl)boronic acid
SMILES CCOC(=O)C1=C(C=C(Cl)C=C1)B(O)O

Specifications

Melting Point 106°C to 108°C
Quantity 1 g
Formula Weight 228.44
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'