missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid, 98%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Melting Point | 106°C to 108°C |
Quantity | 250 mg |
Formula Weight | 228.44 |
Percent Purity | 98% |
Chemical Name or Material | 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'