missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid, 98%
£43.05 - £123.26
Chemical Identifiers
CAS | 871329-55-4 |
---|---|
Molecular Formula | C9H10BClO4 |
Molecular Weight (g/mol) | 228.44 |
MDL Number | MFCD07363744 |
InChI Key | SGPMCOBGNHZGJJ-UHFFFAOYSA-N |
Synonym | 5-chloro-2-ethoxycarbonyl phenylboronic acid, 5-chloro-2-ethoxycarbonyl phenyl boronic acid, 5-chloro-2-ethoxycarbonyl benzeneboronic acid, benzoic acid,2-borono-4-chloro-, 1-ethyl ester 9ci, ethyl 2-borono-4-chlorobenzoate, acmc-209qg0, 5-chloro-2-ethoxycarbonylphenyl boronic acid, 5-chloranyl-2-ethoxycarbonyl-phenyl boronic acid |
PubChem CID | 44784921 |
IUPAC Name | (5-chloro-2-ethoxycarbonylphenyl)boronic acid |
SMILES | CCOC(=O)C1=C(C=C(Cl)C=C1)B(O)O |
Description
Chemical Identifiers
871329-55-4 | |
228.44 | |
SGPMCOBGNHZGJJ-UHFFFAOYSA-N | |
44784921 | |
CCOC(=O)C1=C(C=C(Cl)C=C1)B(O)O |
C9H10BClO4 | |
MFCD07363744 | |
5-chloro-2-ethoxycarbonyl phenylboronic acid, 5-chloro-2-ethoxycarbonyl phenyl boronic acid, 5-chloro-2-ethoxycarbonyl benzeneboronic acid, benzoic acid,2-borono-4-chloro-, 1-ethyl ester 9ci, ethyl 2-borono-4-chlorobenzoate, acmc-209qg0, 5-chloro-2-ethoxycarbonylphenyl boronic acid, 5-chloranyl-2-ethoxycarbonyl-phenyl boronic acid | |
(5-chloro-2-ethoxycarbonylphenyl)boronic acid |
Specifications
871329-55-4 | |
C9H10BClO4 | |
5-chloro-2-ethoxycarbonyl phenylboronic acid, 5-chloro-2-ethoxycarbonyl phenyl boronic acid, 5-chloro-2-ethoxycarbonyl benzeneboronic acid, benzoic acid,2-borono-4-chloro-, 1-ethyl ester 9ci, ethyl 2-borono-4-chlorobenzoate, acmc-209qg0, 5-chloro-2-ethoxycarbonylphenyl boronic acid, 5-chloranyl-2-ethoxycarbonyl-phenyl boronic acid | |
CCOC(=O)C1=C(C=C(Cl)C=C1)B(O)O | |
228.44 | |
228.44 | |
5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid |
106°C to 108°C | |
MFCD07363744 | |
SGPMCOBGNHZGJJ-UHFFFAOYSA-N | |
(5-chloro-2-ethoxycarbonylphenyl)boronic acid | |
44784921 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures