missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-fluorobenzoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 mL
5 mL
Specifications
Specifications
Density | 1.4590g/mL |
Boiling Point | 103.0°C to 104.0°C (15.0 Torr) |
Flash Point | 100°C |
Assay Percent Range | 96% min. (GC) |
Quantity | 1 mL |
Specific Gravity | 1.459 |
Synonym | 5-chloro-2-fluorobenzoylchloride, 2-fluoro-5-chlorobenzoyl chloride, 5-chloro-2-fluoro-benzoyl chloride, benzoyl chloride, 5-chloro-2-fluoro, acmc-1aho4, buttpark 45\01-10, benzoyl chloride,5-chloro-2-fluoro, 5-chloranyl-2-fluoranyl-benzoyl chloride, 5-chloro-2-fluorobenzoyl chloride |
Formula Weight | 193.01 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-2-fluorobenzoyl chloride |
Safety and Handling
Product Identifier
- 5-Chloro-2-fluorobenzoyl chloride
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'