missing translation for 'onlineSavingsMsg'
Learn More
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine, 97%, Thermo Scientific™ Product Code.: 15436107

5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine, 97%, Thermo Scientific™

Product Code. 15436107
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15436107

Brand: Thermo Scientific Alfa Aesar H33292.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in Suzuki, Stille, Sonogashira, carbonylation, Heck, Buchwald-Hartwig reaction.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1347814-95-2
Molecular Formula C8H6ClNO2
Molecular Weight (g/mol) 183.591
MDL Number MFCD20040113
InChI Key KEGRTUBZRMQESQ-UHFFFAOYSA-N
Synonym 5-chlorofuro 3,2-b pyridin-2-yl methanol, 5-chloro-2-hydroxymethyl furo 3,2-b pyridine
PubChem CID 73995151
IUPAC Name (5-chlorofuro[3,2-b]pyridin-2-yl)methanol
SMILES C1=CC(=NC2=C1OC(=C2)CO)Cl

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Formula Weight 183.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10165371 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10191222 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.